Anti-Troponin I fast skeletal muscle Rabbit Monoclonal Antibody [clone: EPR14926]

Proveedor: Abcam
AB183508-100 AB183508-1ML
ABCAAB183508-100EA 675 EUR
ABCAAB183508-100 ABCAAB183508-1ML
Anti-Troponin I fast skeletal muscle Rabbit Monoclonal Antibody [clone: EPR14926]
Anticuerpos
Rabbit monoclonal [EPR14926] to Troponin I fast skeletal muscle.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Troponin I fast skeletal muscle
Clonality: Monoclonal
Clone: EPR14926
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR