Anti-PSMG1 Rabbit Monoclonal Antibody [clone: EPR10223]

Proveedor: Abcam
AB167396-100 AB167396-1ML
ABCAAB167396-100EA 657 EUR
ABCAAB167396-100 ABCAAB167396-1ML
Anti-PSMG1 Rabbit Monoclonal Antibody [clone: EPR10223]
Anticuerpos
Rabbit monoclonal [EPR10223] to PSMG1.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: PSMG1
Clonality: Monoclonal
Clone: EPR10223
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR