Anti-CD8 beta Rabbit Monoclonal Antibody [clone: EPR26538-16] (Alexa Fluor® 488)

Proveedor: Abcam
AB312909-100UL AB312909-10UL
ABCAAB312909-100ULEA 819 EUR
ABCAAB312909-100UL ABCAAB312909-10UL
Anti-CD8 beta Rabbit Monoclonal Antibody [clone: EPR26538-16] (Alexa Fluor® 488)
Anticuerpos
Alexa Fluor® 488 Rabbit monoclonal [EPR26538-16] to CD8 beta.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD8 beta
Clonality: Monoclonal
Clone: EPR26538-16
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR