Anti-PKC eta Rabbit Monoclonal Antibody [clone: EPR18513]

Proveedor: Abcam
AB179524-100 AB179524-1ML
ABCAAB179524-100EA 707 EUR
ABCAAB179524-100 ABCAAB179524-1ML
Anti-PKC eta Rabbit Monoclonal Antibody [clone: EPR18513]
Anticuerpos
Rabbit monoclonal [EPR18513] to PKC eta.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: PKC eta
Clonality: Monoclonal
Clone: EPR18513
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR