Anti-Cleaved Caspase-3 Rabbit Monoclonal Antibody [clone: EPR19676-17]
Proveedor: Abcam
AB244909-1 AB244909-100
ABCAAB244909-1EA 9320 EUR
ABCAAB244909-1 ABCAAB244909-100
Anti-Cleaved Caspase-3 Rabbit Monoclonal Antibody [clone: EPR19676-17]
Anticuerpos
Rabbit monoclonal [EPR19676-17] to Cleaved Caspase-3 - BSA and Azide free (Detector).
Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Cleaved Caspase-3
Clonality: Monoclonal
Clone: EPR19676-17
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human