Anti-beta III Tubulin Rabbit Monoclonal Antibody [clone: EPR19591]

Proveedor: Abcam
AB238454-1 AB238454-100
ABCAAB238454-1EA 9160 EUR
ABCAAB238454-1 ABCAAB238454-100
Anti-beta III Tubulin Rabbit Monoclonal Antibody [clone: EPR19591]
Anticuerpos
Rabbit monoclonal [EPR19591] to beta III Tubulin - BSA and Azide free.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: beta III Tubulin
Clonality: Monoclonal
Clone: EPR19591
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR