Anti-IRBIT Rabbit Monoclonal Antibody [clone: EPR13409]

Proveedor: Abcam
AB178693-100 AB178693-1ML
ABCAAB178693-100EA 738 EUR
ABCAAB178693-100 ABCAAB178693-1ML
Anti-IRBIT Rabbit Monoclonal Antibody [clone: EPR13409]
Anticuerpos
Rabbit monoclonal [EPR13409] to IRBIT.

Precaución: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: IRBIT
Clonality: Monoclonal
Clone: EPR13409
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR