Anti-Caspase-3 p12 Rabbit Monoclonal Antibody [clone: EPR16888]

Proveedor: Abcam
AB179517-100 AB179517-20UL AB179517-1ML
ABCAAB179517-100EA 663 EUR
ABCAAB179517-100 ABCAAB179517-20UL ABCAAB179517-1ML
Anti-Caspase-3 p12 Rabbit Monoclonal Antibody [clone: EPR16888]
Anticuerpos
Rabbit monoclonal [EPR16888] to Caspase-3 p12.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Caspase-3 p12
Clonality: Monoclonal
Clone: EPR16888
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR