Anti-IP3 receptor Rabbit Monoclonal Antibody [clone: EPR4537]

Proveedor: Abcam
AB239933-100 AB239933-1
ABCAAB239933-100EA 1020 EUR
ABCAAB239933-100 ABCAAB239933-1
Anti-IP3 receptor Rabbit Monoclonal Antibody [clone: EPR4537]
Anticuerpos
Rabbit monoclonal [EPR4537] to IP3 receptor - BSA and Azide free.

Precaución: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: IP3 receptor
Clonality: Monoclonal
Clone: EPR4537
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR