Anti-SARS-CoV-2 Spike RBD Human Monoclonal Antibody [clone: CV30]

Proveedor: Abcam
AB277515-1 AB277515-100
ABCAAB277515-1EA 9160 EUR
ABCAAB277515-1 ABCAAB277515-100
Anti-SARS-CoV-2 Spike RBD Human Monoclonal Antibody [clone: CV30]
Anticuerpos
Human monoclonal [CV30] to SARS-CoV-2 Spike RBD - BSA and Azide free.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: SARS-CoV-2 Spike RBD
Clonality: Monoclonal
Clone: CV30
Conjugation:
Epitope:
Host: Human
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR