Anti-MIP-3 beta/CCL19 Rabbit Monoclonal Antibody [clone: EPR7043(2)]

Proveedor: Abcam
AB192871-100 AB192871-1ML
ABCAAB192871-100EA 675 EUR
ABCAAB192871-100 ABCAAB192871-1ML
Anti-MIP-3 beta/CCL19 Rabbit Monoclonal Antibody [clone: EPR7043(2)]
Anticuerpos
Rabbit monoclonal [EPR7043(2)] to MIP-3 beta/CCL19.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: MIP-3 beta/CCL19
Clonality: Monoclonal
Clone: EPR7043(2)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR