Anti-Kv1.1 potassium channel Mouse Monoclonal Antibody [clone: K36/15 H1L1]

Proveedor: Abcam
AB300662-1 AB300662-100
ABCAAB300662-1EA 9160 EUR
ABCAAB300662-1 ABCAAB300662-100
Anti-Kv1.1 potassium channel Mouse Monoclonal Antibody [clone: K36/15 H1L1]
Anticuerpos
Mouse monoclonal [K36/15] to Kv1.1 potassium channel - BSA and Azide free.

Precaución: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Kv1.1 potassium channel
Clonality: Monoclonal
Clone: K36/15 H1L1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR