Anti-PSMD9 Rabbit Monoclonal Antibody [clone: EPR12947]

Proveedor: Abcam
AB250563-100 AB250563-1
ABCAAB250563-100EA 1020 EUR
ABCAAB250563-100 ABCAAB250563-1
Anti-PSMD9 Rabbit Monoclonal Antibody [clone: EPR12947]
Anticuerpos
Rabbit monoclonal [EPR12947] to PSMD9 - BSA and Azide free.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: PSMD9
Clonality: Monoclonal
Clone: EPR12947
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR