Anti-CAPON/NOS1AP Rabbit Monoclonal Antibody [clone: EPR14519(B)]

Proveedor: Abcam
AB190686-100 AB190686-1ML
ABCAAB190686-100EA 650 EUR
ABCAAB190686-100 ABCAAB190686-1ML
Anti-CAPON/NOS1AP Rabbit Monoclonal Antibody [clone: EPR14519(B)]
Anticuerpos
Rabbit monoclonal [EPR14519(B)] to CAPON/NOS1AP.

Precaución: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CAPON/NOS1AP
Clonality: Monoclonal
Clone: EPR14519(B)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR