Anti-RNA polymerase II CTD repeat YSPTSPS (phospho S5) Rabbit Monoclonal Antibody [clone: EP1510Y]

Proveedor: Abcam
AB247458-100 AB247458-1
ABCAAB247458-100EA 1020 EUR
ABCAAB247458-100 ABCAAB247458-1
Anti-RNA polymerase II CTD repeat YSPTSPS (phospho S5) Rabbit Monoclonal Antibody [clone: EP1510Y]
Anticuerpos
Rabbit monoclonal [EP1510Y] to RNA polymerase II CTD repeat YSPTSPS (phospho S5) - BSA and Azide free.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: RNA polymerase II CTD repeat YSPTSPS
Clonality: Monoclonal
Clone: EP1510Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR