Anti-Histone H4 (acetyl K16) Rabbit Monoclonal Antibody [clone: EPR1004]

Proveedor: Abcam
AB194352-100 AB194352-1
ABCAAB194352-100EA 1020 EUR
ABCAAB194352-100 ABCAAB194352-1
Anti-Histone H4 (acetyl K16) Rabbit Monoclonal Antibody [clone: EPR1004]
Anticuerpos
Rabbit monoclonal [EPR1004] to Histone H4 (acetyl K16) - BSA and Azide free.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Histone H4
Clonality: Monoclonal
Clone: EPR1004
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR