Anti-beta IV Tubulin Rabbit Monoclonal Antibody [clone: EPR16775]

Proveedor: Abcam
AB179504-100 AB179504-1ML
ABCAAB179504-100EA 744 EUR
ABCAAB179504-100 ABCAAB179504-1ML
Anti-beta IV Tubulin Rabbit Monoclonal Antibody [clone: EPR16775]
Anticuerpos
Rabbit monoclonal [EPR16775] to beta IV Tubulin.

Type: Primary
Antigen: beta IV Tubulin
Clonality: Monoclonal
Clone: EPR16775
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR