Anti-Uroplakin III Rabbit Monoclonal Antibody [clone: EPR14420]
Proveedor: Abcam
AB239006-100 AB239006-1
ABCAAB239006-100EA 1020 EUR
ABCAAB239006-100 ABCAAB239006-1
Anti-Uroplakin III Rabbit Monoclonal Antibody [clone: EPR14420]
Anticuerpos
Rabbit monoclonal [EPR14420] to Uroplakin III - BSA and Azide free.
Precaución: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Uroplakin III
Clonality: Monoclonal
Clone: EPR14420
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human