Anti-GSK3 beta Mouse Monoclonal Antibody [clone: 7/GSK-3b]
Proveedor: Abcam
AB280381-100 AB280381-1
ABCAAB280381-100EA 1020 EUR
ABCAAB280381-100 ABCAAB280381-1
Anti-GSK3 beta Mouse Monoclonal Antibody [clone: 7/GSK-3b]
Anticuerpos
Mouse monoclonal [7/GSK-3b] to GSK3 beta - BSA and Azide free.
Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: GSK3 beta
Clonality: Monoclonal
Clone: 7/GSK-3b
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse;Rat