Anti-Dopamine beta Hydroxylase Rabbit Monoclonal Antibody [clone: EPR20385]

Proveedor: Abcam
AB209487-100 AB209487-1ML
ABCAAB209487-100EA 682 EUR
ABCAAB209487-100 ABCAAB209487-1ML
Anti-Dopamine beta Hydroxylase Rabbit Monoclonal Antibody [clone: EPR20385]
Anticuerpos
Rabbit monoclonal [EPR20385] to Dopamine beta Hydroxylase.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Dopamine beta Hydroxylase
Clonality: Monoclonal
Clone: EPR20385
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR