Anti-SPINK1/P12 Rabbit Monoclonal Antibody [clone: SP166]

Proveedor: Abcam
AB271943-100 AB271943-1
ABCAAB271943-100EA 1020 EUR
ABCAAB271943-100 ABCAAB271943-1
Anti-SPINK1/P12 Rabbit Monoclonal Antibody [clone: SP166]
Anticuerpos
Rabbit monoclonal [SP166] to SPINK1/P12 - BSA and Azide free.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: SPINK1/P12
Clonality: Monoclonal
Clone: SP166
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR