Anti-PKC delta (phospho Y311) Rabbit Monoclonal Antibody [clone: EPR2609Y]

Proveedor: Abcam
AB247449-100 AB247449-1
ABCAAB247449-100EA 1020 EUR
ABCAAB247449-100 ABCAAB247449-1
Anti-PKC delta (phospho Y311) Rabbit Monoclonal Antibody [clone: EPR2609Y]
Anticuerpos
Rabbit monoclonal [EPR2609Y] to PKC delta (phospho Y311) - BSA and Azide free.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: PKC delta
Clonality: Monoclonal
Clone: EPR2609Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR