Anti-SEMA4A Rabbit Monoclonal Antibody [clone: EPR27097-20]
Proveedor: Abcam
AB314648-20UL AB314648-100UL AB314648-1ML
ABCAAB314648-20ULEA 244 EUR
ABCAAB314648-20UL ABCAAB314648-100UL ABCAAB314648-1ML
Anti-SEMA4A Rabbit Monoclonal Antibody [clone: EPR27097-20]
Anticuerpos
Rabbit monoclonal [EPR27097-20] to SEMA4A.
Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: SEMA4A
Clonality: Monoclonal
Clone: EPR27097-20
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human