Anti-IL-18 Rabbit Monoclonal Antibody [clone: EPR19954-188] (Biotin)
Proveedor: Abcam
AB316302-10UL AB316302-100UL
ABCAAB316302-10ULEA 238 EUR
ABCAAB316302-10UL ABCAAB316302-100UL
Anti-IL-18 Rabbit Monoclonal Antibody [clone: EPR19954-188] (Biotin)
Anticuerpos
Biotin Rabbit monoclonal [EPR19954-188] to IL-18.
Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: IL-18
Clonality: Monoclonal
Clone: EPR19954-188
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human