Anti-N6-methyladenosine (m6A) Rabbit Monoclonal Antibody [clone: EPXVJ-1-2]

Proveedor: Abcam
AB314476-20UL AB314476-100UL AB314476-1ML
ABCAAB314476-20ULEA 0 EUR
ABCAAB314476-20UL ABCAAB314476-100UL ABCAAB314476-1ML
Anti-N6-methyladenosine (m6A) Rabbit Monoclonal Antibody [clone: EPXVJ-1-2]
Anticuerpos
Rabbit monoclonal [EPXVJ-1-2] to N6-methyladenosine (m6A).

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: N6-methyladenosine (m6A)
Clonality: Monoclonal
Clone: EPXVJ-1-2
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR