Anti-Tyrosine Hydroxylase Chicken Monoclonal Antibody [clone: EP1532Y]

Proveedor: Abcam
AB318307-100UG AB318307-1MG
ABCAAB318307-100UGEA 1020 EUR
ABCAAB318307-100UG ABCAAB318307-1MG
Anti-Tyrosine Hydroxylase Chicken Monoclonal Antibody [clone: EP1532Y]
Anticuerpos
Chicken monoclonal [EP1532Y] to Tyrosine Hydroxylase - Chimeric – BSA and Azide free.

Precaución: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Tyrosine Hydroxylase
Clonality: Monoclonal
Clone: EP1532Y
Conjugation:
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR