Anti-POLE Rabbit Monoclonal Antibody [clone: EPR28640-56]

Proveedor: Abcam
AB318137-100UG AB318137-1MG
ABCAAB318137-100UGEA 1020 EUR
ABCAAB318137-100UG ABCAAB318137-1MG
Anti-POLE Rabbit Monoclonal Antibody [clone: EPR28640-56]
Anticuerpos
Rabbit monoclonal [EPR28640-56] to POLE (mutated P286R) - BSA and Azide free.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: POLE
Clonality: Monoclonal
Clone: EPR28640-56
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR