Anti-Histidine decarboxylase Rabbit Monoclonal Antibody [clone: EPR26392-79]

Proveedor: Abcam
AB315445-1MG AB315445-100UG
ABCAAB315445-1MGEA 9160 EUR
ABCAAB315445-1MG ABCAAB315445-100UG
Anti-Histidine decarboxylase Rabbit Monoclonal Antibody [clone: EPR26392-79]
Anticuerpos
Rabbit monoclonal [EPR26392-79] to Histidine decarboxylase - BSA and Azide free.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Histidine decarboxylase
Clonality: Monoclonal
Clone: EPR26392-79
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR