Anti-Influenza A Polymerase basic protein 2 Rabbit Monoclonal Antibody [clone: EPR29356-84]
Proveedor: Abcam
AB320836-100UG AB320836-1MG
ABCAAB320836-100UGEA 1020 EUR
ABCAAB320836-100UG ABCAAB320836-1MG
Anti-Influenza A Polymerase basic protein 2 Rabbit Monoclonal Antibody [clone: EPR29356-84]
Anticuerpos
Rabbit monoclonal [EPR29356-84] to Influenza A Polymerase basic protein 2 - BSA and Azide free.
Precaución: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Influenza A Polymerase basic protein 2
Clonality: Monoclonal
Clone: EPR29356-84
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now