Anti-Clostridium difficile Toxins A + B Rabbit Monoclonal Antibody [clone: EPR23358-136]

Proveedor: Abcam
AB315116-1MG AB315116-100UG
ABCAAB315116-1MGEA 9200 EUR
ABCAAB315116-1MG ABCAAB315116-100UG
Anti-Clostridium difficile Toxins A + B Rabbit Monoclonal Antibody [clone: EPR23358-136]
Anticuerpos
Rabbit monoclonal [EPR23358-136] to Clostridium difficile Toxins A + B - BSA and Azide free (Capture).

Precaución: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Clostridium difficile Toxins A + B
Clonality: Monoclonal
Clone: EPR23358-136
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Clostridium difficile
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR