Anti-Complement Factor B (Ba fragment) Rabbit Monoclonal Antibody [clone: EPR25032-130]

Proveedor: Abcam
AB316818-1MG AB316818-100UG
ABCAAB316818-1MGEA 9200 EUR
ABCAAB316818-1MG ABCAAB316818-100UG
Anti-Complement Factor B (Ba fragment) Rabbit Monoclonal Antibody [clone: EPR25032-130]
Anticuerpos
Rabbit monoclonal [EPR25032-130] to Complement Factor B (Ba fragment) - BSA and Azide free (Capture).

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Complement Factor B (Ba fragment)
Clonality: Monoclonal
Clone: EPR25032-130
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR