Anti-53BP1 (phospho S25) Rabbit Monoclonal Antibody [clone: EPR27057-33]
Proveedor: Abcam
AB315015-20UL AB315015-100UL AB315015-1ML
ABCAAB315015-20ULEA 244 EUR
ABCAAB315015-20UL ABCAAB315015-100UL ABCAAB315015-1ML
Anti-53BP1 (phospho S25) Rabbit Monoclonal Antibody [clone: EPR27057-33]
Anticuerpos
Rabbit monoclonal [EPR27057-33] to 53BP1 (phospho S25).
Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: 53BP1
Clonality: Monoclonal
Clone: EPR27057-33
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human