Anti-SQSTM1 / p62 Rabbit Recombinant Multiclonal Antibody [clone: RM1079]
Proveedor: Abcam
AB314504-20UL AB314504-100UL AB314504-1ML
ABCAAB314504-20ULEA 207 EUR
ABCAAB314504-20UL ABCAAB314504-100UL ABCAAB314504-1ML
Anti-SQSTM1 / p62 Rabbit Recombinant Multiclonal Antibody [clone: RM1079]
Anticuerpos
Rabbit recombinant multiclonal [RM1079] to SQSTM1 / p62.
Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: SQSTM1 / p62
Clonality: Recombinant Multiclonal
Clone: RM1079
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat