Anti-Hexokinase Type III/HK3 Rabbit Monoclonal Antibody [clone: EPR29196-29]
Proveedor: Abcam
AB321996-100UL AB321996-1ML
ABCAAB321996-100ULEA 694 EUR
ABCAAB321996-100UL ABCAAB321996-1ML
Anti-Hexokinase Type III/HK3 Rabbit Monoclonal Antibody [clone: EPR29196-29]
Anticuerpos
Rabbit monoclonal [EPR29196-29] to Hexokinase Type III/HK3.
Precaución: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Hexokinase Type III/HK3
Clonality: Monoclonal
Clone: EPR29196-29
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse