Anti-MBD4/MED1 Rabbit Monoclonal Antibody [clone: EPR28263-16]
Proveedor: Abcam
AB315462-20UL AB315462-100UL AB315462-1ML
ABCAAB315462-20ULEA 275 EUR
ABCAAB315462-20UL ABCAAB315462-100UL ABCAAB315462-1ML
Anti-MBD4/MED1 Rabbit Monoclonal Antibody [clone: EPR28263-16]
Anticuerpos
Rabbit monoclonal [EPR28263-16] to MBD4/MED1.
Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: MBD4/MED1
Clonality: Monoclonal
Clone: EPR28263-16
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human