Anti-T-bet/Tbx21 Rabbit Monoclonal Antibody [clone: EPR27094-16] (Alexa Fluor® 647)

Proveedor: Abcam
AB315957-10UL AB315957-100UL
ABCAAB315957-10ULEA 238 EUR
ABCAAB315957-10UL ABCAAB315957-100UL
Anti-T-bet/Tbx21 Rabbit Monoclonal Antibody [clone: EPR27094-16] (Alexa Fluor® 647)
Anticuerpos
Alexa Fluor® 647 Rabbit monoclonal [EPR27094-16] to T-bet/Tbx21.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: T-bet/Tbx21
Clonality: Monoclonal
Clone: EPR27094-16
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR