unipur Assembly Gloves, 60943 Series
Proveedor: UVEX
6094306 6094307 6094308 6094309 6094310
UVEX6094306EA 25.1 EUR
UVEX6094306 UVEX6094307 UVEX6094308 UVEX6094309 UVEX6094310
unipur Assembly Gloves, 60943 Series
Guantes
The uvex unipur is suitable for mechanical precision work in dry or slightly damp working environments.
  • Polyurethane coating on palm and fingertips
  • Outstanding tactile feel
  • Highly flexible
The robust PU coating is very abrasion-resistant, while still providing outstanding tactile sensation.