Mouse Procollagen III N-terminal propeptide (PIIINP) ELISA Kits
Proveedor: Abcam
AB320800-10X96 AB320800-1X384 AB320800-96TST
ABCAAB320800-10X96EA 8160 EUR
ABCAAB320800-10X96 ABCAAB320800-1X384 ABCAAB320800-96TST
Mouse Procollagen III N-terminal propeptide (PIIINP) ELISA Kits
Ensayos ELISAs
Mouse Procollagen III N-terminal propeptide (PIIINP) ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse Procollagen III N-terminal propeptide (PIIINP) with a sensitivity of 11,21 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10×96 plates for bulk orders and 384-well plate for higher throughput.