Durapore® Millipak® Final Fill Capsules

Proveedor: Merck
MFGL10GF3 MFGL20GF3
MILXMFGL10GF3EA 0 EUR
MILXMFGL10GF3 MILXMFGL20GF3
Durapore® Millipak® Final Fill Capsules
Filtros Filtros de membranas
  • Directions for Use: Organism Retention: Microorganism

  • Mode of Action: Filtration (size exclusion)

  • Application: BioProcessing

  • Intended Use: Reduction or removal of microorganism/bioburden

  • Instructions for Use: Please refer general guidelines section of user guide shipped with this product

  • Storage Statement: Please refer user guide shipped with this product

  • Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations


Material: PVDF membrane
polysulfone housing
silicone O-ring

Fitting: fitting (19 mm sanitary flange inlet and outlet)


Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR