Vacuum concentrators, refrigerated, CentriVap®
Proveedor: LABCONCO
7310030 7310031 7310035 7310036 7310032 7310034
531-0189EA 16330 EUR
531-0189 531-0190 LBCP7310035 531-0998 531-1030 LBCP7310034
Vacuum concentrators, refrigerated, CentriVap®
Evaporadores Evaporadores centrífugos
The refrigerated CentriVap centrifugal concentrator is designed to rapidly concentrate multiple small heat-sensitive samples, such as RNA and proteins, using centrifugal force, vacuum and controlled temperature. The concentrator combines a refrigeration system that cools to −4 °C with a heater that warms to 100 °C to achieve the user's desired set point temperatures. With an epoxy-coated aluminum chamber.
Vacuum pumps provide a reliable vacuum source for the concentrators.
CentriVap Rotors hold tubes at an angle to maximize the surface area and are easily interchangeable. Glassware isMore Product Information