VWR® Degassers, PTFE AF
Proveedor: COLLECTION PREFIX HPLC
1-6353A 0003-6623 0001-6624 0003-6626 0003-6625
903-0134EA 0 EUR
903-0134 903-0156 903-0157 HPLC0003-6626 HPLC0003-6625
VWR® Degassers, PTFE AF
Desgasificantes
On-Line degassing has been shown to be more effective at removing dissolved gas from mobile phases than sonication or helium sparging. Up to five solvent lines can be degassed simultaneously by one unit. The extremely low volume of each PTFE AF channel (480 μl) offers very quick equilibration and very short start-up times.
  • Tiempos de equilibrado y reequilibrado considerablemente más cortos
  • Muy fácil de cebar
  • Tiempos cortos de bajada de vacío; normalmente, 3 minutos
  • El PTFE AF tiene la capacidad de eliminar burbujas antes de la bomba
Bomba de vacíoMore Product Information