Heated product shelf chambers for FreeZone® freeze dryer
Proveedor: LABCONCO
780801030 780801130
LBCP780801030EA 12310 EUR
LBCP780801030 LBCP780801130
Heated product shelf chambers for FreeZone® freeze dryer
Mobiliario Estantes
Heated product shelves with clear chamber for drying bulk samples, or drying serum bottles, vials or microplates. Chamber connects to a FreeZone® freeze dryers for control and recording of the shelf temperature from the Lyo-Works display.
  • Wth removable 254 mm Ø shelves
  • Each shelf is height adjustable to accommodate a broad variety of sample sizes
  • Shelf temperature may be set from –20 to 60 °C
  • Shelves may be placed in freezer for pre-freezing
Each chamber includes a 305 mm Ø clear acrylic cylinder, vacuum release valve and 203 mm Ø opening to collectorMore Product Information