CPC® Miniature Quick-Disconnect Fittings, Panel-Mount Hose Barb Bodies
Proveedor: Avantor Fluid Handling
06363-68 06363-68BLK 06363-97BLK 06363-76BLK
MFLX06363-68EA 60.6 EUR
MFLX06363-68 MFLX06363-68BLK MFLX06363-97BLK MFLX06363-76BLK
CPC® Miniature Quick-Disconnect Fittings, Panel-Mount Hose Barb Bodies
Racores y conectores
No more kinked tubing - allows tubing to rotate freely.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Twist-to-connect couplings provide a reliable and more secure alternative to luer-type connections
  • Tubing can rotate freely when connected - preventing kinked tubing and accidental disconnection
These CPC® miniature quick-disconnect couplings are able to handle ¹/₁₆" ID tubing, and offer a more secure connection than luer fittings. Fitting halves engage and seal with a twist. Once engaged, the coupling allows the tubing to rotateMore Product Information