Anti-BMS-931699 Recombinant Antibody

Proveedor: ProSci Inc.
10-663
PRSI10-663EA 654 EUR
PRSI10-663
Anti-BMS-931699 Recombinant Antibody
Anticuerpos
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: (scFv-heavy-kappa)-(scFv-heavy-lambda)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR