Anti-BAY 1213790 Recombinant Antibody

Proveedor: ProSci Inc.
10-916
PRSI10-916EA 654 EUR
PRSI10-916
Anti-BAY 1213790 Recombinant Antibody
Anticuerpos
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1-kappa-lambda
Reactivity:
New Product
Order Now

ESPECIFICACIONES

Environmentally Preferable
Sinónimos de antígenos BCD-100
Tipo de anticuerpo Primario
Clonalidad Recombinant
Conjugado Unconjugated
Reactividad
Huésped
Isotipo
Modificación None
Inmunógeno Homo sapiens,Chimeric/PDCD1/PD1/CD279 [Homo sapiens]
Purification >95%
Concentración Batch dependent
Storage buffer PBS buffer pH 7,5
Temperatura de almacenamiento Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2…8 °C for 1 - 2 weeks. Store at –20 °C for 12 months. Store at –80 °C for long term storage.
Shipping temperature 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR