Anti-ITGAX Recombinant Antibody [Clone: N418]

Proveedor: ProSci Inc.
24-370
PRSI24-370EA 737 EUR
PRSI24-370
Anti-ITGAX Recombinant Antibody [Clone: N418]
Anticuerpos
Type: Primary
Antigen: integrin subunit alpha X
Clonality: Recombinant
Clone: N418
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Mouse;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR