Anti-CD154 Recombinant Antibody [Clone: YMF323.6]

Proveedor: ProSci Inc.
24-529
PRSI24-529EA 737 EUR
PRSI24-529
Anti-CD154 Recombinant Antibody [Clone: YMF323.6]
Anticuerpos
Type: Primary
Antigen: CD154
Clonality: Recombinant
Clone: YMF323.6
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR