Anti-Cd40lg Recombinant Antibody [Clone: MR1]

Proveedor: ProSci Inc.
24-623
PRSI24-623EA 737 EUR
PRSI24-623
Anti-Cd40lg Recombinant Antibody [Clone: MR1]
Anticuerpos
Type: Primary
Antigen: CD40 ligand
Clonality: Recombinant
Clone: MR1
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR