Anti-EIF3H Rabbit Polyclonal Antibody

Proveedor: ProSci Inc.
30-620
PRSI30-620EA 660 EUR
PRSI30-620
Anti-EIF3H Rabbit Polyclonal Antibody
Anticuerpos
EIF3H belongs to the peptidase M67 family. It contains 1 MPN (JAB/Mov34) domain. EIF3H binds to the 40S ribosome and promotes the binding of methionyl-tRNAi and mRNA. It associates with the p170 subunit of EIF3.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: EIF3H
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR