Anti-ORF62 Mouse Monoclonal Antibody
Proveedor: ProSci Inc.
32-229
PRSI32-229EA 509 EUR
PRSI32-229
Anti-ORF62 Mouse Monoclonal Antibody
Anticuerpos
Kaposi's sarcoma-associated herpesvirus (KSHV) belongs to the gamma-(2)-herpesvirus subfamily and has been closely linked to the Kaposi’s sarcoma, primary effusion lymphoma (PEL) and multicentric Castleman’s disease. The genome of KSHV is 165-170 kb and contains at least 88 open reading frames. At least five major proteins are likely to be involved in the assembly of the HHV-8 capsid, including a protease (encoded by ORF17), the major capsid protein (encoded by ORF25), and three other smaller capsid proteins (encoded by ORF62, ORF26, and ORF65). Previous structural studies have shown thatMore Product Information